Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Heavy Duty Orbital Shaker, 230 V

Supplier: Eberbach
E6155.25
75805-252EA 49105.85 USD InStock
75805-252
Heavy Duty Orbital Shaker, 230 V
Shakers and Mixers Orbital Shakers
One of the highest capacity orbital shakers on the market. With ability to shake up to 500 lbs of sample. Eberbach’s proprietary closed loop controller ensures the most accurate mixer speeds at all rpms; if accuracy and repeatability is what you require Eberbach has the best in class! Also, featuring a built in pallet jack base for easy transportation and placement.

  • 230 V model
  • 500 lbs. weight capacity
  • 30 to 300 rpm
  • Digital controller
  • 5 year warranty

Eberbach’s large capacity Variable-Speed Orbital Shakers are designed for rotary swirling action and is used for bioprocessing of plant cell cultures, large batch mammalian cell culturing, aeration of fermentations and various other chemical mixing procedures.

The orbital motion of the E6155 ensures efficient liquid mixing. The E6155 allows high oxygen transfer rates with low shear forces not seen with other bio-reactors which use propellers or impellers. The E6155 reduces stress on cells thanks to the swirling action of the shaker. This motion is particularly suitable for cell cultivation and fermentation. The E6155 uses Eberbach unique balanced bearing plates for a long lasting smooth operation, repeatable motion with accurate control of the orbital speed and time duration.

When used with optional biochemical sensors, the E6155 is ideal for scale-up fermentation, cultivation and bio processing applications. The base is designed to be moved with a forklift or pallet jack. The E6155 uses Eberbach’s own controller which has a digital tachometer along with digital countdown timer and optional audible end of cycle alarm. The digital countdown timer can be selected for timed operation or turned off for constant operation.  The eccentric drive is counterbalanced to assure smooth acceleration and minimal bearing wear for increased life of the machine.  Speed is continuously variable between from 30 - 300 rpm or with the Eberbach controller you can program your specific speed. The E6155 comes factory set with a 2” (5 cm) orbit, if your application requires a different orbit size we can offer alternative sizes.

Accessories information: The E6155 can be supplied with multiple optional features; remote control box mounted to a pendant with castors or as a wall mount box, Sanitary models (stainless steel with IP67 rated motor), hazardous location models (explosion-proof), custom top platforms and more. Contact your Avantor representative for more information.

Ordering information: Box Carrier is not included (Box Carrier E6155.A.UB pictured). Accessories not included.

Packaging: Product ships LTL in a crate.

Caution: 230 V model. Corresponding model with voltage suitable for use in US (75805-250) is also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty