Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HisCube HisCube Ni-Indigo His-Tag Protein Purification MINI Kit

Supplier: Cube Biotech
HisCube
80101
77393-512EA 760 USD InStock
77393-512
HisCube HisCube Ni-Indigo His-Tag Protein Purification MINI Kit
Protein Purification Protein Purification Reagents
The HisCube His-tag protein purification MINI Kit is our way to supply your laboratory with a convenient method for high-quality, small-scale His-tag protein purifications. We put all of our experience in His-tag purifications into this Kit to provide a protocol that is easy to follow.

  • Usage: Specific binding and purification of 6× His tagged proteins
  • Specificity: Affinity to His-tagged proteins
  • Binding capacity:>100 mg/ml
  • Bead Ligand: INDIGO-Ni
  • Bead size: 90 μm
  • Filling quantity: Delivered as a 50% suspension
  • pH stability: 2 - 14
  • Chelator stability (e.g. EDTA): 20 mM
  • Reducing agent stability (e.g. DTT): 20 mM
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile

Features - MagBead separator:
Usage: Separation of magnetic beads from the supernatant
Separation of magnetic beads from the supernatant: 1.5 and 2 ml microcentrifuge tubes (Eppis)
Design: Modular (2 microcentrifuges tubes/module): Can be put together onto a larger rack easily
Magnet strength: Above average. To ensure no magnetic beads get lost during the separation process
Area covered on work bench: 3 cm² per module
Cap holder: Included, so that microcentrifuge tubes do not fall close if not wanted
Color: White

The HisCube Ni-INDIGO MINI kit contains our in-house developed PureCube 100 Ni-INDIGO agarose resin and our PureCube 1-Step batch MINI batch spin tubes.

INDIGO's most significant advantage is its tolerance towards reducing and chelating agents. These can be used to great help in His-tag purifications, but traditional His-tag affinity beads do not tolerate them. EDTA is a chelator often used in eukaryotic cell buffers because it has protease-inhibiting functions. DTT can dissolve protein aggregates that could hinder the access of the purification beads to the His-tagged proteins. However, thanks to INDIGO, both substances can be used now. Therefore, they are part of the kit.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty