Would you like to log in again?
BRAND® QuickSip™ BT-Aspirator
Supplier: BrandTech
BRAND® QuickSip™
4723180
77296-294EA 293.36 USD InStock
77296-294
BRAND® QuickSip™ BT-Aspirator
Pumps Vacuum Aspiration Systems
Safely and quickly aspirate up to 25 ml of common lab liquids per pump, no vacuums or power needed.
  • Safe removal of supernatants
  • Works without vacuum pump
  • Obtain fingertip vacuum control using the cell-culture™ hand controller asapiration unit (available separately)
  • Suitable for use with disposable pipette tips, micropipette tips, and pasteur pipettes
  • Up to 25 ml per plunger stroke
  • Optional eight-channel dispensing manifold is also available separately
  • Non-autoclavable
The QuikSip™ BT-aspirator is a simple bottle-top solution for liquid aspiration in the lab. Designed for aspirating aqueous solutions and various organic and inorganic substances, it offers versatility for a range of applications, including biological buffers and cell culture liquids.
Setting up QuikSip™ is easy: just attach it to bottles with 45 mm, S40, or 33 mm threads, then press down the top to activate. Liquid is collected directly into the bottle (bottle not included).
To ensure optimal performance and safety, it's important to check compatibility with specific liquids.
Delivery information: Includes QuikSip™ bottle-top aspirator, spare dispensing cartridge, two PP adapters (GL45/32 and FL45/S40).
Caution: Avoid using QuikSip™ with substances that may damage materials like PP, PE, or EPDM, such as non-polar solvents or concentrated acids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty