Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PureCube Amine Activated Agarose XL

Supplier: Cube Biotech
PureCube
59005 59010 59012
77393-406EA 6517 USD InStock
77393-406 77393-404 77393-402
PureCube Amine Activated Agarose XL
Protein Purification Protein Purification Reagents
PureCube amine activated agarose XL resin is your best option to couple biomolecules carrying free-standing carboxy (COOH) groups.

  • Usage: Coupling proteins to active Amine Groups
  • Specificity: Exposed carboxy (COOH) groups on biomolecule
  • Amine group density: Higher than 20 µmol/ml
  • Bead ligand: Chemically active amine groups
  • Bead size: 400 μm
  • Filling quantity: Delivered as a 50% suspension
  • Storage buffer: 100% isopropanol
  • Matrix: 7.5% cross-linked agarose
  • Short term storage: In neutral buffer at 4 °C
  • Long term storage: 100 mM sodium hydrogen carbonate, 0.02% sodium azide, pH 7.5 at 4 °C or 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
  • Shelf life: If properly stored - three years upon delivery, unopened
  • Chemical Structure: Amine structure

The resulting peptide bond between the Amine Agarose beads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube Amine Activated Agarose is also available as magnetic beads in case you prefer this purification method, or as agarose resin with smaller beads if you need beads with higher mechanical stability.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty