Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Decyl-β-D-glucopyranoside ≥99.5%, Crystallization grade

Supplier: CUBE BIOTECH
Synonyms: Decyl glucopyranosideDGN-Decyl beta-D-glucopyranosideDecyl beta-D-glucopyranoside

16062 16061 16060
77392-394PK 1513 USD InStock
77392-394 77392-396 77392-392
Decyl-β-D-glucopyranoside ≥99.5%, Crystallization grade
Decyl β-D-glucopyranoside
Mild, non-ionic detergents such as Decylglucoside are well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer glucosides with chain lengths from C-8 to C-10, as well as octylthioglucoside.

Molecular Weight: 306.40 g/mol
Form: White crystalline powder
CMC (in H2O): 2.2 mM


Formula: C₁₆H₃₂O₆
MW: 320.43 g/mol
MDL Number: MFCD00063297
CAS Number: 58846-77-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty