Would you like to log in again?
Vacuum/Pressure Pumps Coated with PTFE
Supplier: Cole Parmer
EW-79200-35
MSPP-EW7920035EA 544.51 USD InStock
MSPP-EW7920035
Vacuum/Pressure Pumps Coated with PTFE
Pumps Vacuum Pumps
Wetted parts are coated with PTFE to handle your toughest applications.
  • Lightweight aluminum construction and compact size
  • Pumps with gauges and regulators give you more control of your vacuum/pressure applications
These pumps are ideal for benchtop applications such as vacuum filtration. Pumps include suction cup feet and a 6-ft power cord with built-in switch (the 115 VAC models have grounded plug, 220 VAC and DC models have stripped ends).
Ordering information: Three years warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty