Would you like to log in again?
Sartobind® Luer Adapters
Supplier: Sartorius
Sartobind®
17002140 1ZA0005 1ZA0005 17002140
76795-402EA 57.75 USD InStock
CA76795-402 76795-402 CA76795-400 76795-400
Sartobind® Luer Adapters
Fittings and Connectors
Adapters to convert the Luer outlet connector on any Sartobind® Lab unit to the UNF 10-32 standard. This enables the operation of Sartobind® Lab units on FPLC systems fitted with UNF 10-32 male connectors.
  • Adapters for connecting Sartobind® Lab units to most liquid chromatography systems
  • Easily convert female Luer lock inlet connectors to the UNF 10-32 standard
  • Tool-free, fingertight connection
This is a replacement part - a minimum of one piece is already provided in each box of Sartobind® Lab unit(s).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty