Would you like to log in again?
Cryo Tube Storage Box, PC, 100-Place
Supplier: Heathrow Scientific
HS23300D
77395-382PK 46.2 USD InStock
77395-382
Cryo Tube Storage Box, PC, 100-Place
Boxes Cryoboxes
Safely store cryogenic vials.
  • Our polycarbonate boxes are designed with unique features to help orient, identify and access vials with ease
  • Alphanumeric labeling on lid or base (depending on the unit) for quick location and retrieval of tubes
  • Liquid nitrogen vent and drainage holes
  • Magnet on top of forceps enables easy storing on metal surfaces
  • Stackable, saves valuable space in the lab
The Arctic Squares® polycarbonate cryostorage box safely stores vials from −196 to +121 °C in mechanical freezers as well as liquid nitrogen. These polycarbonate boxes are designed with unique features to help orient, identify and access vials with ease.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty