Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ampicillin sodium salt for molecular biology

Supplier: IBI Scientific
Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt

IB02040
IB02040EA 103.79 USD InStock
IB02040
Ampicillin sodium salt for molecular biology
Ampicillin sodium salt


Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39
MDL Number: MFCD00064313
CAS Number: 69-52-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty