Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Rubbermaid® Commercial Food/Tote Box Lids, Essendant
Supplier: Essendant
Rubbermaid®
RCP3302CLE RCP3302CLE RCP3310CLE RCP3502WHI RCP3510WHI
62660-620EA 81.3 USD InStock
62660-620 62660-825 62660-429 62660-824 62660-949
Rubbermaid® Commercial Food/Tote Box Lids, Essendant
Foodservice Supplies
This bulk food storage container lid reduces risk of cross-contamination, secures storage while stacked, and can be used as tray when displayed upside down.
  • Crystal-clear material provides visibility, durability and stain-resistance
  • Tight-fitting, snap-on lid with stacking ribs keeps food secure
  • Date control panels for easy inventory control
The Rubbermaid Commercial food storage lid for food tote box helps reduce food spoilage costs.
Certifications: FDA Compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty