Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4'-Bipyridine -1,1'-dioxide 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4,4'-Bipyridine N,N'-dioxide[4,4'-Bipyridine] 1,1'-dioxide

OR22806-1G OR22806-5G OR22806-10G
77406-954EA 124 USD InStock
77406-954 77406-952 77406-950
4,4'-Bipyridine -1,1'-dioxide 98%
4,4'-Bipyridine -1,1'-dioxide


Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
Melting Pt: 298-306 °C
MDL Number: MFCD00134383
CAS Number: 24573-15-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty