Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Adamantanethiol 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 1-MercaptoadamantaneAdamantane-1-thiol

OR309163-25G OR309163-5G
77408-990EA 277 USD InStock
77408-990 77408-992
1-Adamantanethiol 98%
1-Adamantanethiol


Formula: C₁₀H₁₆S
MW: 168.298 g/mol
Melting Pt: 103-105 °C
MDL Number: MFCD00213530
CAS Number: 34301-54-7
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty