Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dihydroxybenzoic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: beta-Resorcylic acid

OR4893-1KG OR4893-100G OR4893-500G
77422-252EA 20 USD InStock
77422-252 77422-254 77422-256
2,4-Dihydroxybenzoic acid 97%
2,4-Dihydroxybenzoic acid


Formula: C₇H₆O₄
MW: 154.12 g/mol
Melting Pt: 208-218 °C
MDL Number: MFCD00002451
CAS Number: 89-86-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty