Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Bromoheptanoic acid 94%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 7-Bromoenanthic Acid

OR62069-25G OR62069-100G OR62069-5G
77410-788EA 162 USD InStock
77410-788 77410-792 77410-790
7-Bromoheptanoic acid 94%
7-Bromoheptanoic acid
Radiopharmaceutical precursor


Formula: C₇H₁₃BrO₂
MW: 209.08 g/mol
MDL Number: MFCD00037800
CAS Number: 30515-28-7
UN: 3261
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty