Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanolamine 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Monoethanolamine2-Aminoethanol

OR921532-100G
77411-750EA 20 USD InStock
77411-750
Ethanolamine 99%
Ethanolamine


Formula: NH₂CH₂CH₂OH
MW: 61.08 g/mol
Boiling Pt: 170 °C
Melting Pt: 10-11 °C
Density: 1.012
Flash Pt: 91 °C
MDL Number: MFCD00008183
CAS Number: 141-43-5
UN: 2491
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty