Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Epichlorhydrin 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 1-Chloro-2,3-epoxypropane(±)-Glycerol epichlorhydrinEpichlorohydrin(±)-1-Chloro-2,3-epoxypropane(±)-3-Chloro-1,2-epoxypropane3-Chloro-1,2-epoxypropaneEpichlorhydrin

OR922424-25G OR922424-500G
77411-802EA 59 USD InStock
77411-802 77411-804
(±)-Epichlorhydrin 99%
(±)-Epichlorhydrin


Formula: C₃H₅ClO
MW: 92.52 g/mol
Boiling Pt: 115-117 °C
Melting Pt: -57 °C
Density: 1.183
Flash Pt: 28 °C
MDL Number: MFCD00005132
CAS Number: 106-89-8
UN: 2023
ADR: 6.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty