Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(1-Propynyl)tributyltin 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Tributyl(1-propynyl)tin

OR961474-5G OR961474-1G OR961474-25G
77412-234EA 750 USD InStock
77412-234 77412-232 77412-236
(1-Propynyl)tributyltin 98%
(1-Propynyl)tributyltin


Formula: C₁₅H₃₀Sn
MW: 329.12 g/mol
Boiling Pt: 277 °C
Density: 1.082
Flash Pt: > 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863650
CAS Number: 64099-82-7
UN: 2788
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty