Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium sulfide 20.0 - 24.0% in aqueous solution, AR®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = N

MACR3524-04 MACR3524-04
MK352404CS 3342.83 USD InStock
MK352404 CAMK352404
Ammonium sulfide 20.0 - 24.0% in aqueous solution, AR®, Macron Fine Chemicals™
Ammonium sulphide


Formula: H₈N₂S
MW: 68.14 g/mol
MDL Number: MFCD00010892
CAS Number: 12135-76-1
EINECS: 235-223-4
UN: 2683
ADR: 8 (6.1) (3),II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (as (NH₄)₂S) 20.0 - 24.0 %
Antimony (Sb) ≤0.0005 %
Arsenic (As) ≤0.0001 %
Carbonate (CO₃) ≤0.005 %
Chloride (Cl) ≤0.005 %
Residue after Ignition ≤0.04 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty