Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trihexylamine 96%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Tri-n-hexylamineTri(hex-1-yl)amine

OR10061-50G OR10061-10G OR10061-250G
77402-438EA 96 USD InStock
77402-438 77402-436 77402-434
Trihexylamine 96%
Trihexylamine


Formula: C₁₈H₃₉N
MW: 269.51 g/mol
Boiling Pt: 263…265 °C
Density: 0.794 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009523
CAS Number: 102-86-3
EINECS: 203-062-9
UN: 3082
ADR: 9+EHS,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty