Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroiodomethane 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR30688-100G OR30688-25G OR30688-500G
77419-022EA 16 USD InStock
77419-022 77419-020 77419-024
Chloroiodomethane 98%
Chloroiodomethane


Formula: CH₂ClI
MW: 176.38 g/mol
Boiling Pt: 108…109 °C
Density: 2.422 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001078
CAS Number: 593-71-5
EINECS: 209-804-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty