Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Hydroxypicolinic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 5-hydroxypyridine-2-carboxylic acid

OR5545-1G OR5545-5G
77423-082EA 21 USD InStock
77423-082 77423-080
5-Hydroxypicolinic acid 95%
5-Hydroxypicolinic acid


Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 260…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00661309
CAS Number: 15069-92-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty