Would you like to log in again?
Accucold® Vaccine Refrigerators with Removable Ventilated Drawers, 2 to 8 °C
Supplier: Felix Storch Inc
Accucold®
ARG61PVBIADADR ARS62PVBIADADR
77434-712EA 3889 USD InStock
77434-712 77434-704
Accucold® Vaccine Refrigerators with Removable Ventilated Drawers, 2 to 8 °C
Refrigerators
Refrigerators for display and refrigeration of vaccines, units have removable ventilated drawers, ideal for pharmacy, medication, and vaccine applications.
  • Sized just under 32" high to fit under lower ADA compliant counters
  • Intelligent microprocessor digital temperature controller
  • Improved temperature accuracy with a separate temperature probe for continuous product readout to simulate stored product temperature
  • Three removable drawers allow easy access, movement and storage of interior contents
  • Recessed installation capable for use in 24" wide spaces
Accucold's Pharma-Vac Performance Series are upright all-refrigerators designed and purpose-built for pharmacy, medication and vaccine applications to support meeting CDC/VFC guidelines.
These models are 6 cu.ft. all-refrigerator with a temperature range of 2 to 8 °C. Sized at just under 32" high to fit under lower counters complying with ADA guidelines for accessibility. They are designed for freestanding or recessed installation. The microprocessor temperature controller is externally located to minimize door openings, helping to protect stored contents from ambient temperature. The buffered temperature probe is encased in a glycol-filled bottle to better simulate temperature of the stored product, with an additional sensor that reads air temperature to ensure superior temperature control. The current and min/max temperature display is viewable in Celsius or Fahrenheit to the nearest tenth of a degree. The audible and visual temperature alarm sounds if the unit's interior goes out of range. Additional alarms include a power failure alert, door open alarm, and sensor failure alarm. These models are equipped with remote alarm contacts in the rear and a ½" probe port on the right side to accept additional monitoring devices. A hospital grade cord with 'green dot' plug is also included for added safety in high traffic areas.
Inside, the ARG61PVBIADADR utilizes adjustable cycle defrost operation to avoid temperature spikes. Optimized forced air cooling with an internal fan to ensure excellent temperature stability and uniformity with rapid recovery after door openings. Three white powder-coated aluminum drawers provide ample storage and easy organization. Positioned on ball-bearing glides, the drawers offer full-extension for complete product access. Additionally, each drawer is removable allowing for transport of contents as needed. The drawers also lock into place for added security, while the perforated bottom ensures proper ventilation throughout the unit. This unit includes LED internal lighting with an on/off rocker switch. The interior is constructed from white powder coated metal to ensure added durability and better temperature retention.
The ARG61PVBIADADR comes in a white cabinet finish with a blue accented control panel. The factory-installed antimicrobial handle is powder-coated with naturally occurring silver ions, which help to reduce the spread of germs, allowing for a safer, cleaner user experience. The double pane tempered glass door provides a full view of the interior. The user-reversible door ships with a right hand swing and self-closing function, while a factory-installed lock provides security as needed, with two keys for staff use. Powerful magnetic gaskets ensure a positive seal.
Designed with hydrocarbon refrigerant, the ARG61PVBIADADR provides an energy efficient solution to vaccine storage. This unit is Pyxis, Omnicell, and AcuDose RX compatible. Additional sizes, as well as solid door options, are also available.
Caution: Accucold refrigerators are purpose-built to the support meeting the current CDC (Centers for Disease Control) guidelines for the storage of vaccines. Units that end in suffix "456" have been certified to the new NSF 456 standard. As referenced in the current Vaccine Storage and Handling Toolkit, these units incorporate microprocessor controls and forced air circulation for stable operation between 2 and 8 °C. For additional temperature assurance, the CDC recommends a calibrated digital data logger (DDL), which can be added with the factory-installed option DL2B or addition of the DL2BKit accessory.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty