Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetone ≥99.7%, OmniSolv® HR-GC for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: 2-PropanoneDMKPropanoneDimethyl ketone

AX0110-1 AX0110-1
EM-AX0110-1CS 1636 USD InStock
CAAX0110-1 EM-AX0110-1
Acetone ≥99.7%, OmniSolv® HR-GC for gas chromatography, Supelco®
Acetone
For chromatography, residue analysis, and spectrophotometry. Filtered through 0.2µm filter. Lot analysis and chromatograms on label.
Impurities by Capillary GC:


Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/07/720607.jpg
Order Now
Specification Test Results
Assay (GC) 99.7% min
Appearance Clear liquid, free from particulates
Cap. ECD responsive substances as PCNB (peak) 2ng/L max
Cap. ECD responsive substances as PCNB (total) 50ng/L max
Cap. FID responsive substances as decane (peak) 1µg/L max
Cap. FID responsive substances as decane (total) 10µg/L max
Color (APHA) 5 max
Filtered through 0.2 µm filter To pass test
Residue after evaporation 1ppm max
Titrable acid 0.3µeq/g max
Water (H2O) 0.5% max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty