Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.9%, ChromAR® HPLC super gradient, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Methane, cyano-Methyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

MACR2856-10 MACR2856-10
MK285610CS 2169.82 USD InStock
CAMK285610 MK285610
Acetonitrile ≥99.9%, ChromAR® HPLC super gradient, Macron Fine Chemicals™
Acetonitrile
ChromAR—Solvents that meet ACS specifications and are suitable for liquid chromatography and UV-spectrophotometry. 


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (CH₃CN) ≥ 99.9 %
Water (KF) ≤ 0.02 %
Titrable Acid (meq/g) ≤ 0.0005
Titrable Base (meq/g) ≤ 0.0002
Residue after Evaporation ≤ 0.0005%
Ultraviolet Absorbance (1.00-cm cell vs. water):
195 mm ≤ 0.12
200 nm ≤ 0.03
210 nm ≤ 0.02
220 nm ≤ 0.01
254 - 400 nm ≤ 0.005
Gradient Elution Test 210 nm ≤ 3 mAU
Filtered through a 0.2 micron filter.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty