Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Butanol ≥98.0%, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: sec-Butyl alcohol(±)-2-Hydroxy butane(±)-sec-Butyl alcohol(±)-sec-Butanolsec-Butanol(±)-Methyl ethyl carbinol(±)-Ethyl methyl carbinol

BX1795-6 BX1795-6
EM-BX1795-6CS 850 USD InStock
EM-BX1795-6 CABX1795-6
(±)-2-Butanol ≥98.0%, Supelco®
(±)-2-Butanol


Formula: H₃CCH₂CH(OH)CH₃
MW: 74.12 g/mol
Boiling Pt: 94 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0.8534 g/cm³ (20 °C)
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00004569
CAS Number: 78-92-2
EINECS: 201-158-5
UN: 1120
ADR: 3,III
Merck Index: 13,01540
https://us-legacy.vwr.com/stibo/web/std.lang.all/03/13/710313.jpg
Order Now
Specification Test Results
Assay (GC) 98.0% min
Color (APHA) 20 max
Form Clear liquid
Infrared spectrum Conforms to standard

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty