Would you like to log in again?
Accucold® Commercial Chest Freezers with Digital Thermostat, −45 °C
Supplier: Felix Storch Inc
Accucold®
EL51LT EL41LT EL21LT EL31LT EL11LT
77434-714EA 3889 USD InStock
77434-714 77434-716 77434-718 77434-720 77434-722
Accucold® Commercial Chest Freezers with Digital Thermostat, −45 °C
Freezers Laboratory / General Purpose Freezers
Commercial −45 °C capable chest freezers with digital thermostat for laboratory and industrial storage purposes.
  • Capable of −45 °C operation for select laboratory and industrial storage purposes
  • UL-S listed to NSF-7 commercial standards
  • Digital thermostat with external controls allows easy and precise temperature monitoring
  • Factory installed lock for security you can depend on
  • Extra thick 4½" insulation for efficient and stable cooling
Accucold EL models are −45 °C capable chest freezers built with solid insulation for laboratory and life science storage applications.
These chest freezers use static manual defrost cooling to maintain constant temperature, with an externally controlled digital thermostat to ensure easy temperature monitoring. Extra thick 4½" thick insulation offers added efficiency and reliability. Additional features include a solid, easy-grip handle complete with factory installed lock, a sturdy set of casters, and counterbalanced hinges for comfortable loading and unloading. The units includes a factory-installed probe hole to allow users to install their own monitoring equipment.
The models are UL-S listed to ANSI-NSF standards for commercial use and is designed to operate between −10 and −45 °C. Additional sizes are available. Please contact your Avantor sales representative for more options.
Caution: Accucold freezers are purpose-built to the support meeting the current CDC (Centers for Disease Control) guidelines for the storage of vaccines. As referenced in the current Vaccine Storage and Handling Toolkit, these units incorporate several key features for stable operation. For additional temperature assurance, the CDC recommends a calibrated digital data logger (DDL), which can be added with the factory-installed option DL2B or addition of the DL2BKit accessory. Select Accucold products have been tested and certified to the new NSF 456 standard, and are now available for purchase. For additional information, contact our products specialists.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty