Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Buffered oxide etch (6:1), CMOS for microelectronic use, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = R

1178-03 BAKR1178-03
JT1178-3CS 591.34 USD InStock
CAJT1178-3 JT1178-3
Buffered oxide etch (6:1), CMOS for microelectronic use, J.T.Baker®
Buffered oxide etch
In the Avantor Electronic Materials range, we have solvents, acids and bases in CMOS and Finyte/VLSI quality, as well as Performance Materials like photoresist- and residue-removers, selective etchants, BEOL and FEOL. Avantor Performance Materials portfolio of production-proven chemistries, customizable chemistry platforms and world-class applications centers are designed to help you meet your process improvement goals faster and speed up your ramp time to the next node.


UN: 2992
ADR: 8,II
Order Now
Specification Test Results
For Microelectronic Use
Ship and store material above 60°F (16°C)
Ammonium Fluoride (NH₄F)(w/w) 33.5 - 35.5 %
Hydrofluoric Acid (HF)(w/w) 7.0 - 7.3 %
Residue after Ignition ≤10 ppm
Chloride (Cl) ≤3000.0 ppb
Nitrate (NO₃) ≤3000 ppb
Phosphate (PO₄) ≤700.0 ppb
Sulfate (SO₄) ≤1500.0 ppb
Trace Impurities - Aluminum (Al) ≤50.0 ppb
Arsenic and Antimony (as As) ≤30.0 ppb
Trace Impurities - Barium (Ba) ≤10.0 ppb
Trace Impurities - Beryllium (Be) ≤10.0 ppb
Trace Impurities - Bismuth (Bi) ≤20.0 ppb
Trace Impurities - Boron (B) ≤50 ppb
Trace Impurities - Cadmium (Cd) ≤20.0 ppb
Trace Impurities - Calcium (Ca) ≤100.0 ppb
Trace Impurities - Chromium (Cr) ≤10.0 ppb
Trace Impurities - Cobalt (Co) ≤10.0 ppb
Trace Impurities - Copper (Cu) ≤10.0 ppb
Trace Impurities - Gallium (Ga) ≤10.0 ppb
Trace Impurities - Germanium (Ge) ≤40.0 ppb
Trace Impurities - Gold (Au) ≤20 ppb
Heavy Metals (as Pb) ≤1000.0 ppb
Trace Impurities - Iron (Fe) ≤50.0 ppb
Trace Impurities - Lead (Pb) ≤10.0 ppb
Trace Impurities - Lithium (Li) ≤10.0 ppb
Trace Impurities - Magnesium (Mg) ≤50.0 ppb
Trace Impurities - Manganese (Mn) ≤10.0 ppb
Trace Impurities - Molybdenum (Mo) ≤10.0 ppb
Trace Impurities - Nickel (Ni) ≤10.0 ppb
Trace Impurities - Potassium (K) ≤50 ppb
Trace Impurities - Silver (Ag) ≤50 ppb
Trace Impurities - Sodium (Na) ≤100.0 ppb
Trace Impurities - Strontium (Sr) ≤10.0 ppb
Trace Impurities - Thallium (Tl) ≤20.0 ppb
Trace Impurities - Tin (Sn) ≤20.0 ppb
Trace Impurities - Titanium (Ti) ≤100.0 ppb
Trace Impurities - Zinc (Zn) ≤100.0 ppb
Particle Count - 0.5 µm and greater ≤150 par/ml
Particle Count - 1.0 µm and greater ≤8 par/ml
For additional information, go to www.askavantor.com. Search keywords
freezing" and product name."

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty