Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethylene glycol monobutyl ether, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Butyldigol2-(2-Butoxyethoxy)ethanolDiethylene glycol mono-n-butyl ether

Management of Change (MOC) category = N

BAKRD654-09 BAKRD654-09
JTD654-9CS 1611.4 USD InStock
CAJTD654-9 JTD654-9
Diethylene glycol monobutyl ether, J.T.Baker®
Diethylene glycol monobutyl ether


Formula: CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
MW: 162.23 g/mol
Boiling Pt: 230.4 °C (1013 hPa)
Melting Pt: –68 °C
Density: 0.954 g/cm³ (20 °C)
Flash Pt: 78.00 °C
Storage Temperature: Ambient
MDL Number: MFCD00002881
CAS Number: 112-34-5
EINECS: 203-961-6
Merck Index: 12,01592
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty