Would you like to log in again?
Accucold® Laboratory Chest Freezers, −30 or −35 °C
Supplier: Felix Storch Inc
Accucold®
VT73 VT133 VLT204 VT183 VLT74 VLT104 VLT134 VT203 VLT184 VT252 VLT253 VT103
77434-746EA 3377 USD InStock
77434-746 77434-734 77434-756 77434-748 77434-736 77434-738 77434-762 77434-740 77434-750 77434-752 77434-732 77434-754
Accucold® Laboratory Chest Freezers, −30 or −35 °C
Freezers Laboratory / General Purpose Freezers
Laboratory chest freezers with stainless steel corner guards and NIST calibrated thermometer, ideal for laboratory, pharmacy and industrial storage.
  • Designed to operate at −30 or −35 °C depending on model for select laboratory, pharmacy, industrial and other institutional storage needs
  • Includes an external NIST calibrated readout of the current and high/low temperature with an audible alarm
  • Manual defrost operation and static cooling system
  • Lock included for optional security
  • Stainless steel bumpers to protect the freezer's corners
Accucold's low temperature laboratory series includes a range of chest freezers capable of operating at −30 or −35 °C (−22 or 31 °F) depending on model.
The models are manual defrost chest freezers designed to store laboratory and institutional materials. Units includes a digital thermostat and high/low temperature alarm with an external temperature readout, calibrated to NIST standards in our ISO/IEC 17025:2017 accredited laboratory. A two year certificate of calibration is included for your records, as well as the option of re-calibrating your temperature probe at the time of its expiration.
The freezers comes in a white finish with stainless steel bumpers pre-installed on the front corners for added protection in high traffic settings. The counterbalanced hinges allow you to keep the lid open when loading and unloading, with an easy grab handle and keyed lock included. The hammered aluminum interior includes two wire storage baskets.
Additional sizes in this series are available, as well as lower temperature options. Please contact your Avantor sales representative for details to meet your cold storage equipment needs.
Caution: Accucold freezers are purpose-built to the support meeting the current CDC (Centers for Disease Control) guidelines for the storage of vaccines. As referenced in the current Vaccine Storage and Handling Toolkit, these units incorporate several key features for stable operation. For additional temperature assurance, the CDC recommends a calibrated digital data logger (DDL), which can be added with the factory-installed option DL2B or addition of the DL2BKit accessory. Select Accucold products have been tested and certified to the new NSF 456 standard, and are now available for purchase. For additional information, contact our products specialists.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty