Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Babcock Cream Test Bottle Brush, Mill-Rose
Supplier: Mill-Rose
72220B
17065-000PK 43.12 USD InStock
17065-000
Babcock Cream Test Bottle Brush, Mill-Rose
Brushes
Special construction puts the cleaning action directly where it is needed.
  • Along the brush stem, rows of short dense bristles provide extra scrubbing for the neck
  • All natural black bristles twisted into a wire handle with a hanging loop
The extra long radial bristles fold to fit through the narrow bottle neck then expand to fill the chamber.
Neck diameter: 1.3 cm (1/2")
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty