Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Hepatitis C Virus RNA-directed RNA polymerase Rabbit Polyclonal Antibody

Supplier: Bioss

BS-4858R
77437-100EA 375 USD InStock
77437-100
Anti-Hepatitis C Virus RNA-directed RNA polymerase Rabbit Polyclonal Antibody
Antibodies
The RNA directed RNA polymerase is also known as non-structural protein NS5B. NS5B is a 65 kDa protein that resembles other viral RNA polymerases. Hepatitis C virus (HCV) replication is thought to occur in membrane bound replication complexes. These complexes transcribe the positive strand and the resulting minus strand is used as a template for the synthesis of genomic RNA. There are two viral proteins involved in the reaction, NS3 and NS5B.

Type: Primary
Antigen: Hepatitis C Virus RNA-directed RNA polymerase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Virus

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty