Would you like to log in again?
VWR® PTFE Stoppers
Supplier: Avantor
VWR®
011.478.03 011.478.07 011.478.05 011.478.06 011.478.04 011.478.08 011.478.02
77441-910CS 1316 USD InStock
77441-910 77441-912 77441-908 77441-914 77441-904 77441-916 77441-906
VWR® PTFE Stoppers
Stoppers PE/LDPE/HDPE Stoppers
Solid PTFE stoppers with T-shaped head.
  • T-handles for easy on and off
For use with volumetric flasks and separating funnels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty