Would you like to log in again?
Accucold® Ultra Low Temperature Chest Freezers, −50 to −85 °C
Supplier: Felix Storch Inc
Accucold®
LAB8R6 LAB8 LAB11 LAB11R9
77434-724EA 14365 USD InStock
77434-724 77434-726 77434-728 77434-730
Accucold® Ultra Low Temperature Chest Freezers, −50 to −85 °C
Freezers Ultra Low Temperature Freezers
ULT chest freezers for laboratory and life science storage of biological samples, chemicals select vaccines and antibiotics.
  • Ideal for industrial research and other materials with strict temperature requirements
  • Extra thick 4½" insulation for efficient and stable cooling
  • Digital thermostat with external controls allows easy and precise temperature monitoring
  • Factory installed lock for security you can depend on
  • Counterbalanced hinges allow easier loading
Accucold's series of ultra low temperature chest freezers have been designed for reliable storage at −85 °C for laboratory, life science and other institutional settings.
Extra thick 4½" insulation and static manual defrost operation help to maintain ultra low temperatures suitable for maintaining the integrity of biological samples, industrial materials, laboratory specimens, select vaccines and other materials with highly specific cold storage requirements. Two inner lids offer added protection by shielding contents when the outer lid is opened. The digital microprocessor controls are mounted in the front for convenient access. The units have a temperature range that can be set between −50 and −85 °C, with a display viewable in Celsius or Fahrenheit. The thermostat's high/low temperature alarm warns if the interior goes out of the set range. There is also an audible 9 V battery-powered alarm in the rear of the unit that will sound in the event of power failure.
For added convenience, the LAB11 comes with pre-installed casters, including two front locking wheels to help keep the unit in place. The easy-grip handle includes a factory-installed keyed lock for added security. Choice of unit with a set of 9 vertical stainless steel box racks (see model LAB11R9).
Certifications: UL and UL-C listed, approved for use in USA and Canada, approved to ANSI/NSF-7.
Ordering information: LAB8R6 units are supplied with 3× 10-box shelves and 3× 5-box shelves in stainless steel and the LAB11R9 units are supplied with 6× 10-box shelves and 3× 5-box shelves in stainless steel for added storage convenience.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty