Would you like to log in again?
XTRA-PFE™ Latex Examination Gloves, Kimberly-Clark
Supplier: Kimberly-Clark
KIMBERLY-CLARK® XTRA-PFE®
50503 50501 50504 50502 50502 50501 50503
32934-122CS 395.67 USD InStock
32934-122 CA32934-122 32934-124 CA32934-124 32934-126 CA32934-126 32934-128
XTRA-PFE™ Latex Examination Gloves, Kimberly-Clark
Gloves
These powder-free gloves offer outstanding barrier protection from exposure to blood, bodily fluids, and certain chemicals.
  • Ambidextrous
  • Beaded cuff
  • For single use only
  • AQL 1.5
  • Thickness: 10.2 mil (finger)
Fully textured for a secure grip, these gloves are 50% thicker and 20% longer than standard examination gloves. Tested for use with 10 commonly used chemotherapy drugs to determine permeation and breakthrough times.
Caution: This product contains natural rubber latex, which may cause allergic reactions. Safe use of this glove by or on latex-sensitive individuals has not been established. This product should not be worn by or exposed to individuals allergic to natural rubber latex.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty