Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Best Master Natural Rubber Gloves, Showa

Supplier: Showa

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

709M-08 709XL-10 709S-07 709L-09
32892-075CS 656.22 USD InStock
32892-075 32892-078 32892-077 32892-076
Best Master Natural Rubber Gloves, Showa
Gloves
Hand and product protection for use in chemical plants, asbestos handling, janitorial, general construction, fiberglass work, automotive assembly, canning, electronics, and maintenance.

  • Acceptable for food and pharmaceutical applications
  • Embossed for excellent wet/dry grip

Constructed of 100% premium quality natural rubber latex. Chlorinated for additional case-hardened chemical resistance to animal fats, caustics, acids, oils, salts, alcohol, and detergents. Gloves provide tactile sensitivity, good tensile strength, and tear/puncture resistance. Color: yellow.

Certifications: Gloves are made using only listed materials and levels outlined in 21 CFR 170-199.

Ordering information: Pairs are individually poly bagged.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty