Would you like to log in again?
Comfort Flex® Flock-Lined Latex Gloves, Magid
Supplier: Magid
Comfort Flex®
620XL 620L 620S 620M
69100-037PK 26.71 USD InStock
69100-037 69100-039 69100-089 69100-041
Comfort Flex® Flock-Lined Latex Gloves, Magid
Gloves
General-purpose, hand-specific rubber latex gloves with flock lining, pinked cuff edges, and embossed grip.
  • Embossed finish for fantastic grip
  • Stands up well to everyday chemicals
  • Comes on and off with ease
This is fully liquid-proof, making it perfect when dealing with household chemicals, as well as certain mild abrasives.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty