Would you like to log in again?
Chloride Color Reagent, for Chloride Determination by the Automated Ferricyanide Method, Ricca Chemical Company
Supplier: RICCA Chemical Company
R1940000-4C R1940000-1C
RC1940-32EA 178.64 USD InStock
RC1940-32 RC19401
Chloride Color Reagent, for Chloride Determination by the Automated Ferricyanide Method, Ricca Chemical Company
Environmental Test Kits
Clear, orange-yellow liquid
For chloride determination by the automated ferricyanide method.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty