Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Dichlorobenzene ≥98%, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: o-Dichlorobenzene

DX0685-3 DX0685-3
EM-DX0685-3EA 824.15 USD InStock
EM-DX0685-3 CADX0685-3
1,2-Dichlorobenzene ≥98%, Supelco®
1,2-Dichlorobenzene


Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 180.5 °C (1013 hPa)
Melting Pt: –17 °C
Density: 1.309 g/cm³ (20 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00000535
CAS Number: 95-50-1
EINECS: 202-425-9
UN: 1591
ADR: 6.1,III
Merck Index: 13,03081
Order Now
Specification Test Results
Assay (GC) 98% min
Color (APHA) 20 max
Form Clear liquid
Infrared spectrum Conforms to standard

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty