Would you like to log in again?
KIMTECH PURE® G5 Nitrile Gloves, Kimberly-Clark
Supplier: Kimberly-Clark
KIMTECH PURE® G5
HC250N 56866 56860 56858 56864 56865 56867 56863 HC450N 56862 56858 56862 56859 56866 56863 56861 56860 56864 56868
32933-754CS 769.23 USD InStock
CA32933-758 CA14214-532 32933-754 CA32933-752 CA14215-738 14215-738 32933-752 CA32933-760 32933-758 CA32933-756 CA14215-734 32933-756 CA32933-754 CA14215-736 CA14215-730 14215-730 14215-734 32933-760 14215-732
KIMTECH PURE® G5 Nitrile Gloves, Kimberly-Clark
Gloves
Ambidextrous, powder-free nitrile gloves contain no latex.
  • Static dissipative in use
  • Beaded cuff for easier donning
Light blue gloves have a bisque finish. White gloves have textured fingertips to facilitate control and grip.
Certifications: Recommended for ISO Class 5 (FED-STD-209E Class 100/M3.5) or higher cleanroom environments. Packaged in a Class 100 Cleanroom. AQL 1.5.
Ordering information: Packaged in a Class 100 cleanroom. Packed 100 per double poly bag with case liner.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty