Would you like to log in again?
Hot Start Taq DNA Polymerase 2X Master Mix
Supplier: G-Biosciences
786-1965 786-1964
76716-700EA 218.37 USD InStock
76716-700 76716-626
Hot Start Taq DNA Polymerase 2X Master Mix
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Hot Start Taq DNA polymerase 2X master mix contains chemically modified hot start Taq DNA Polymerase, dNTPs and optimized buffer with stabilizers.
  • Routine PCR, qPCR and genotyping
High GC content template DNA amplification with GC enhancer
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty