Would you like to log in again?
Hot Start High Fidelity DNA Polymerase
Supplier: G-Biosciences
786-1970 786-1971
76716-710EA 682.96 USD InStock
76716-710 76716-670
Hot Start High Fidelity DNA Polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Hot start high-fidelity DNA Polymerase possesses 5´→3´ polymerase and 3´→5´ exonuclease (proofreading) activities. This Polymerase has the fidelity, sensitivity and processivity with an error rate ~2.8×10² fold lower than Taq DNA polymerase, and significantly lower than other proofreading DNA polymerases.
  • High fidelity PCR and cloning
Long and high GC content template DNA amplification.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty