Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Nitron One® Lightly-Powdered Nitrile Gloves, Microflex®, Ansell
Supplier: Ansell
Nitron One®
NO-123-XL
75881-070CS 2161.87 USD InStock
75881-070
Nitron One® Lightly-Powdered Nitrile Gloves, Microflex®, Ansell
Gloves
Tough enough to handle the most difficult jobs, these gloves are longer for more protection over the wrist.
  • Lightly powdered
  • Beaded cuffs
  • Ambidextrous
  • Fully textured for improved grip
These nitrile gloves are fully textured for enhanced wet or dry gripping. Made with 100% synthetic nitrile. For industrial use only.
Packaging: Packaged 100 gloves per dispenser box.
Caution: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex sensitized individuals has not been established.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty