Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accucold® Built-In Undercounter Laboratory Freezers, −30 or −35 °C

Supplier: Felix Storch Inc
Accucold®
VT65ADA VLT650 VLT650ADA VT65
77434-758EA 2665 USD InStock
77434-758 77434-760 77434-742 77434-744
Accucold® Built-In Undercounter Laboratory Freezers, −30 or −35 °C
Freezers Laboratory / General Purpose Freezers
Built-in undercounter laboratory freezers which are commercially listed to NSF-7 standards.

  • Commercially listed to NSF-7 standards
  • Capable of operating at −30 or −35 °C for long-term food preservation and safe storage of pharmaceuticals
  • Slim width and counter height fits easily where space is tight
  • Precise temperature control with a digital thermostat
  • Factory installed lock provides security you can count on
  • Three slide-out freezer drawers help contents maintain stable temperature, even with an open door

Accucold medical all-freezers offer low temperature storage in a perfectly sized footprint.

These models are upright all-freezer sized with a 24" footprint. The ADA units are 32" high, designed to fit under lower ADA compliant counters. The front-breathing design of all models allows built-in installation, while the fully finished cabinet also allows freestanding use. A factory installed keyed lock offers added security. The digital thermostat is located in the kickplate so that users can manage the temperature without opening the door. A NIST calibrated high/low temperature alarm is also included.

Inside, the units utilize static manual defrost operation to maintain stable temperature. The models can reach temperatures as low as −35 °C (VT65/VT65ADA units −30 °C). Three slide-out basket drawers offer convenient storage while minimizing temperature fluctuations when the door is open.

Certifications: Models are ETL-S approved to NSF-7 standards.

Caution: Accucold freezers are purpose-built to the support meeting the current CDC (Centers for Disease Control) guidelines for the storage of vaccines. As referenced in the current Vaccine Storage and Handling Toolkit, these units incorporate several key features for stable operation. For additional temperature assurance, the CDC recommends a calibrated digital data logger (DDL), which can be added with the factory-installed option DL2B or addition of the DL2BKit accessory. Select Accucold products have been tested and certified to the new NSF 456 standard, and are now available for purchase. For additional information, contact our products specialists.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty