Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethylene glycol monoethyl ether, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: 2-(2-Ethoxyethoxy)ethanol

Management of Change (MOC) category = N

BAKRL216-07 BAKRL216-07
JTL216-7EA 123.9 USD InStock
JTL216-7 CAJTL216-7
Diethylene glycol monoethyl ether, J.T.Baker®
Diethylene glycol monoethyl ether


Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: –80 °C
Density: 0.99 g/cm³ (20 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00002872
CAS Number: 111-90-0
EINECS: 203-919-7
REACH: 01-2119475105-42
Merck Index: 13,01809
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Water (H₂O)(by Karl Fischer titrn) ≤0.2 %
Identification (by IR) Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty