Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanol 50% (v/v) analytical reagent

Supplier: Ricca Chemical
Danger
Synonyms: Ethyl alcohol

R2546500-4A
RC2546.50-1CS 2347.18 USD InStock
RC2546.50-1
Ethanol 50% (v/v) analytical reagent
Ethanol
Clear, colorless liquid. Denatured with 2 to 3% methanol and 2 to 3% 2-propanol.

  • Total alcohol: ~ 50%
  • Ethyl alcohol: 23-50%
  • Denaturant: 1-3% methyl alcohol and 1-3% isopropyl alcohol


Formula: C₂H₅OH
MW: 46.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,III
Merck Index: 13,03795
Order Now
Specification Test Results
Appearance Clear, colorless, alcohol odor
Assay 50.0 ± 1.0 % (v/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty