Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride, crystal disk for spectroscopy, for CO₂ lasers, polished, drilled

Supplier: INTERNATIONAL CRYSTAL LABS MS
Synonyms: HaliteSea saltSaline

0002A-84
100212-706EA 83.6 USD InStock
100212-706
Sodium chloride, crystal disk for spectroscopy, for CO₂ lasers, polished, drilled
Sodium chloride
Quality crystal optics for spectroscopy and CO2 lasers.


Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty