Would you like to log in again?
Corning® Ultra-Low Attachment Culture Flasks, Sterile, Corning
Supplier: Corning
Corning®
3814
89089-878CS 784.69 USD InStock
89089-878
Corning® Ultra-Low Attachment Culture Flasks, Sterile, Corning
Flasks Cell Culture Flasks
The ultra-low attachment surface is a covalently bound hydrogel layer that is hydrophilic and neutrally charged
Flasks are manufactured from optically clear virgin polystyrene and are supplied with lot numbers for ease in traceability. Rectangular flasks feature graduations and canted necks, and are supplied with vented HDPE caps. Sterilized by gamma radiation.
The surface minimizes cell attachment, protein absorption, and enzyme activation. It is ideal for culturing stem cells, as it promotes ebryoid body formation from ES cells. Surfaces are stable, noncytotoxic, biologically inert, and nondegradable.
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty