Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PYREX® Distilling Flask, Vertical Type, Standard Taper 24/40 Joints, 3 Necks, Corning
Supplier: Corning
PYREX®
4950-5L 4950-2L 4950-1L
89090-696CS 938.96 USD InStock
89090-696 89090-864 89091-476
PYREX® Distilling Flask, Vertical Type, Standard Taper 24/40 Joints, 3 Necks, Corning
Flasks Distillation Flasks
PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Central neck and two parallel side necks. Matt white labelling area. Three vertical neck flasks have two side necks with 24/40 outer standard taper joints and a center neck with a 24/40 standard taper joint. All the necks are at the same height.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty