Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cetrimonium bromide ≥98.0%, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: N-Cetyl-N,N,N-trimethylammonium bromideCetyltrimethylammonium bromideHexadecyltrimethylammonium bromide(1-Hexadecyl)trimethylammonium bromideN,N,N-Trimethylhexadecan-1-aminium bromideCTAB

Management of Change (MOC) category = N

BAKRN121-20 BAKRN121-20
JTN121-20CS 10211.57 USD InStock
CAJTN121-20 JTN121-20
Cetrimonium bromide ≥98.0%, BAKER ANALYZED®, J.T.Baker®
Cetrimonium bromide


Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364.45 g/mol
Melting Pt: 237…243 °C
Flash Pt: 244 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011772
CAS Number: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9,III
Merck Index: 13,02034
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₁₉H₄₂BrN) ≥98.0 %
Insoluble Matter ≤0.005 %
Residue after Ignition ≤0.10 %
Appearance Passes Test
Identification (by IR) Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty