Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydroxylamine sulfate ≥99.0%, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Bis(hydroxylammonium) sulphate Hydroxylamine sulphate (2:1)

Management of Change (MOC) category = N

BAKRN646-07 BAKRN646-06
JTN646-6CS 2020.42 USD InStock
JTN646-6 JTN646-7
Hydroxylamine sulfate ≥99.0%, J.T.Baker®
Hydroxylammonium sulphate


Formula: H₈N₂O₆S
MW: 164.14 g/mol
Melting Pt: 170 °C
Density: 1.1 g/cm³ (20 °C)
MDL Number: MFCD00044869
CAS Number: 10039-54-0
EINECS: 233-118-8
UN: 2865
ADR: 8,III
Merck Index: 13,04853
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay ((NH₂OH)₂·H₂SO₄) ≥99.0 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty